missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Potassium Sulfate Dodecahydrate, ≥99.0% (RT), ACS Reagent; Honeywell™ Fluka™

Product Code. 15615930 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15615930 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15615930 Supplier Honeywell-Fluka Supplier No. 60151500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 89°C
CAS 7788-99-0
Color Violet
Physical Form Crystalline
Quantity 500 g
Molecular Formula CrH24KO20S2
Linear Formula CrK(SO4)2 · 12H2O
MDL Number MFCD00149917
UN Number NONH for all modes of transport
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
Molecular Weight (g/mol) 499.386
PubChem CID 24596
Formula Weight 499.4g/mol
Percent Purity ≥99.0% (RT)
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Chromium(III) potassium sulfate dodecahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.