missing translation for 'onlineSavingsMsg'
Learn More

Chromium Potassium Sulfate, Dodecahydrate, MP Biomedicals™

Product Code. 11478272 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478272 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11478272 Supplier MP Biomedicals Supplier No. 0219403105
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chromium potassium sulfate is the potassium double sulfate of chromium.

Chromium alum is used in the tanning of leather. It was also used in gelatine emulsions in photographic film as a hardener.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 89°C (lit.)
CAS 7788-99-0
Physical Form Crystals
Quantity 5 g
Assay Percent Range 98.0-102.0%
Molecular Formula CrH24KO20S2
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
Solubility Information Soluble in water.
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
Molecular Weight (g/mol) 499.386
PubChem CID 24596
Formula Weight 499.4g/mol
Grade ACS
Density 1.89 g/cm3 (at 20°C)
Show More Show Less
compliance-icons
Product Identifier
  • Chromium Potassium Sulfate Dodecahydrate, ACS
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.