missing translation for 'onlineSavingsMsg'
Learn More

Chromosulfuric acid, Thermo Scientific Chemicals

Product Code. 10722961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10722961 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10722961 Supplier Thermo Scientific Acros Supplier No. 295510010
  • ONLINE EXCLUSIVE PRICE
    75.20 €
    73.00 € / 1 L
    Save 2.20 €
    2% Off
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 65272-71-1 | Molecular Formula: Cr2K2O7·H2O4S | Molecular Weight: 200.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65272-71-1
Molecular Formula Cr2K2O7·H2O4S
Molecular Weight (g/mol) 200.237
MDL Number MFCD00010952
InChI Key IFMFTIBWWVBKOC-UHFFFAOYSA-L
Synonym absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
PubChem CID 131664149
IUPAC Name dipotassium;chromium;dihydroxide;dihydrate
SMILES O.O.[OH-].[OH-].[K+].[K+].[Cr]

Specifications

Color Brown to Brown-Red
Boiling Point 330.0°C
Density 1.8400g/mL
Packaging Glass bottle
Specific Gravity 1.84
Quantity 1 L
Physical Form Liquid
Solubility Information Solubility in water: soluble in water (caution ! development of heat)
Chemical Name or Material Chromosulfuric acid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Butylmagnesium chloride, 20 wt% solution in THF/Toluene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofuran, Toluene, Butylmagnesiumchloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.