Learn More
Proteintech ChromoTek Cell Cycle Chromobody™ Plasmid (TagRFP)
Cell Cycle-Chromobody™ fused to TagRFP.
Brand:  Proteintech CCR
Additional Details : Weight : 0.00240kg
Product Code. 17353283
  • 874.00 € / 20µg
Estimated Shipment: 31-05-2024
to see stock.
Description
Description
  • DNA plasmid encoding for anti-PCNA VHH (anti-PCNA Nanobody) fused to fluorescent protein TagRFP
  • PCNA is a cell cycle marker
Specifications
Specifications
20 μg