missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GFP-Booster ATTO488

Product Code. 17303343
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17303343 10 μL 10µL
17313343 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17303343 Supplier Proteintech Supplier No. gba48810
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GFP VHH conjugated to ATTO 488.

  • anti-GFP VHH/ Nanobody conjugated to a fluorescent dye for IF/ microscopy

Specifications

Clonality Monoclonal
Quantity 10 μL
For Use With (Application) Immunofluorescence (IF), Immunohistochemistry (IHC), Immunocytochemistry (ICC)
Conjugate ATTO488
Target Specificity AcGFP, Citrine, CFP, eGFP, eYFP, GFP S65T, pHluorin, sfGFP, mClover (Clover A206K), TagGFP, tagGFP2, Venus, wtGFP, YFP
Content And Storage Shipped at ambient temperature. Upon receipt store at +4™C. Stable for 6 month. Do not freeze. Protect from light.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.