missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek GFP-Trap™ Agarose

Product Code. 17393343
Change view
Click to view available options
Sufficient For:
10 Reactions
100 Reactions
20 Reactions
200 Reactions
400 Reactions
Unit Size:
10 reactions
100 reactions
20 reactions
200 reactions
400 reactions
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
17393343 20 Reactions 20 reactions
17313353 400 Reactions 400 reactions
17303353 200 Reactions 200 reactions
17373343 10 Reactions 10 reactions
17383343 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17393343 Supplier Proteintech Supplier No. gta20
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GFP-Trap™ Agarose is an affinity resin for IP of GFP-fusion proteins.

  • Consists of a GFP Nanobody/ VHH coupled to agarose beads

Specifications

Content And Storage Upon receipt store at 4°C. Do not freeze.
For Use With (Application) IP, CoIP, ChIP, RIP
Target AcGFP, Clover, eGFP, Emerald, GFP, GFP5, GFP Envy, GFP S65T, mGFP, mPhluorin, PA-GFP, Superfolder GFP, TagGFP, TagGFP2, monomeric eGFP A206K, CFP, YFP, Citrine, eCitrine, eYFP, Venus, Ypet, BFP
Label or Dye Agarose
Species Alpaca
Sufficient For 20 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.