Learn More
Proteintech ChromoTek GFP-Trap™ Agarose Kit
Click to view available options
Quantity:
20 Reactions
Unit Size:
20 reactions
Description
- GFP-Trap™ Agarose is an affinity resin for IP of GFP-fusion proteins
- Consists of a GFP Nanobody/ VHH coupled to agarose beads
- The ChromoTek GFP-Trap™ Agarose is a ready-to-use reagent
Specifications
Specifications
| Quantity | 20 Reactions |
| Sufficient For | 20 Reactions |