missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgG1, Fc-specific VHH, Biotinylated

Product Code. 17353483
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17353483 10 μL 10µL
17363483 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17353483 Supplier Proteintech Supplier No. smsG1B110
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Nano-CaptureLigand for mouse IgG1: Fc-specific single domain antibody/VHH [CTK0103], biotinylated.

  • The Nano-CaptureLigand mouse IgG1 captures and immobilizes mouse IgG1 antibodies and Fc-fragments to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
  • Consists of a biotinylated Nanobody that specifically binds to the mouse IgG1 subclass with high affinity and in a site-directed manner

Specifications

Quantity 10 μL
Target Specificity Fc-fragment of Mouse IgG1
Conjugate Biotin
Clonality Monoclonal
Content And Storage Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
For Use With (Application) BLI, SPR, ELISA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.