Learn More
Proteintech ChromoTek Nuclear Actin-Chromobody™ Plasmid (TagGFP)
Nuclear Actin-Chromobody™ fused to TagGFP2.
Brand:  Proteintech ACG-N
Additional Details : Weight : 0.00240kg
Product Code. 17313283
  • 874.00 € / 20µg
Estimated Shipment: 05-06-2024
to see stock.
Description
Description
  • DNA plasmid encoding for anti-Actin VHH (anti-actin Nanobody)with nuclear localization signal (NLS) fused to TagGFP2
Specifications
Specifications
20 μg