Learn More
Proteintech ChromoTek Vimentin-Chromobody™ Plasmid (TagGFP)
Vimentin-Chromobody™ fused to TagGFP2.
Brand:  Proteintech VCG
Additional Details : Weight : 0.00240kg
Product Code. 17333553
  • 874.00 € / 20µg
Estimated Shipment: 05-06-2024
to see stock.
Description
Description
  • DNA plasmid encoding for anti-Vimentin VHH (anti-Vimentin Nanobody) fused to TagGFP2
Specifications
Specifications
20 μg