missing translation for 'onlineSavingsMsg'
Learn More

Chromplete™ Water, HPLC/GC, and Spectrometry

Product Code. 15930721 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
2.5 L
4 L
Unit Size:
1 L
2.5 L
4 L
This item is not returnable. View return policy

Product Code. 15930721

Brand: Thermo Scientific T001032500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Water is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry. | CAS: 7732-18-5 | H2O | 18.02 g/mol

Chromplete Water is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name water
SMILES O

Specifications

Absorbance ≤ 0.01, ≤ 0.005
pH 5.5-7.0
Color ≤5 APHA
CAS Min % 0.99
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.01
Grade Chromplete
Packaging Amber Glass
Residue after Evaporation ≤ 1.0 ppm
Quantity 2.5 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.