missing translation for 'onlineSavingsMsg'
Learn More

Chromplete™ Water, HPLC/GC, and Spectrometry

Product Code. 15940721 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
Unit Size:
1 L
2.5 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15940721 4 L 4 L
15920721 1 L 1 L
15930721 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15940721 Supplier Thermo Scientific Supplier No. T001034000
  • ONLINE EXCLUSIVE PRICE
    61.95 €
    57.50 € / 4 L
    Save 4.45 €
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Water is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry. | CAS: 7732-18-5 | H2O | 18.02 g/mol

Chromplete Water is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name water
SMILES O

Specifications

Absorbance ≤ 0.01, ≤ 0.005
pH 5.5-7.0
Color ≤5 APHA
CAS Min % 0.99
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.01
Grade Chromplete
Packaging Amber Glass
Residue after Evaporation ≤ 1.0 ppm
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.