missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chrysene-d12, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Chrysene D12 |
| CAS | 1719-03-5 |
| Molecular Formula | C18 2H12 |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C18H12/c1-3-7-15-13(5-1)9-11-18-16-8-4-2-6-14(16)10-12-17(15)18/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c([2 H])c([2 H])c3c4c([2 H])c([2 H])c([2 H])c([2 H])c4c([2 H])c([2 H])c23 |
| IUPAC Name | 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriochrysene |
| Molecular Weight (g/mol) | 240.36 |
| Formula Weight | 240.1692 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction