missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CHS 828
GREENER_CHOICE

Product Code. 18758426
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18758426

Brand: Tocris Bioscience™ 6753/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NAMPT inhibitor; active in vivo and cytotoxic

CHS 828 is a NAMPT inhibitor. Decreases cellular level of NAD+. Selectively increases intracellular ROS levels in cancer cells but not normal cells. Active in vivo and cytotoxic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[6-(4-Chlorophenoxy)hexyl]-N'-cyano-N''-4-pyridinylguanidine
CAS 200484-11-3
Quantity 25 mg
Molecular Formula C19H22ClN5O
Formula Weight 371.86
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.