missing translation for 'onlineSavingsMsg'
Learn More

α-Chymotrypsin ≥40u/mg protein, MP Biochemicals™

Product Code. 11413150 Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.1 g
250 mg
500 mg
1 g
Unit Size:
100mg
1g
250mg
500mg
This item is not returnable. View return policy

Product Code. 11413150

Brand: MP Biomedicals 0210046190

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 9004-07-3
MDL Number MFCD00130481

Specifications

Color White
Quantity 500 mg
Physical Form Powder
Chemical Name or Material α-Chymotrypsin, 3X Crystallized from Bovine Pancreas, ≥40μ/mg protein
compliance-icons compliance-icons
Product Identifier
  • Chymotrypsin
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : chymotrypsin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.