missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CI 988

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective CCK2 (CCK-B) receptor antagonist that displays ∽ 1600-fold selectivity over CCK1 receptors (IC50 values are 1.7 and 2717 nM for CCK2 and CCK1 respectively). Has negligible affinity at a range of other binding sites (IC50 > 10 μM). Exhibits anxiolytic activity following oral administration.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 614.73 |
| Purity | >99% |
| Molecular Weight (g/mol) | 614.743 |
| Chemical Name or Material | CI 988 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction