missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CI 988
GREENER_CHOICE

Product Code. 18074202
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18074202

Brand: Tocris Bioscience™ 2607/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective CCK2 antagonist

Potent and selective CCK2 (CCK-B) receptor antagonist that displays ∽ 1600-fold selectivity over CCK1 receptors (IC50 values are 1.7 and 2717 nM for CCK2 and CCK1 respectively). Has negligible affinity at a range of other binding sites (IC50 > 10 μM). Exhibits anxiolytic activity following oral administration.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130332-27-3
Molecular Formula C35H42N4O6

Specifications

Quantity 50 mg
Formula Weight 614.73
Purity >99%
Molecular Weight (g/mol) 614.743
Chemical Name or Material CI 988
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.