missing translation for 'onlineSavingsMsg'
Learn More

CibenzolineSuccinate, TRC

Product Code. 30352804
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352804 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352804 Supplier Toronto Research Chemicals Supplier No. TRCC5357255MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cibenzoline Succinate
CAS 100678-32-8
Molecular Formula C18 H18 N2 . C4 H6 O4
Quantity 5 mg
Synonym Butanedioic acid, compd. with 2-(2,2-diphenylcyclopropyl)-4,5-dihydro-1H-imidazole (1:1), 1H-Imidazole, 2-(2,2-diphenylcyclopropyl)-4,5-dihydro-, butanedioate (1:1) (9CI), Cibenol, Cibenzoline succinate, Cibenzoline succinate (1:1), Cifenline succinate, Cipralan, Exacor, Ro 22-7796/001
InChI Formula InChI=1S/C18H18N2.C4H6O4/c1-3-7-14(8-4-1)18(15-9-5-2-6-10-15)13-16(18)17-19-11-12-20-17;5-3(6)1-2-4(7)8/h1-10,16H,11-13H2,(H,19,20);1-2H2,(H,5,6)(H,7,8)
SMILES OC(=O)CCC(=O)O.C1CN=C(N1)C2CC2(c3ccccc3)c4ccccc4
IUPAC Name butanedioic acid;2-(2,2-diphenylcyclopropyl)-4,5-dihydro-1H-imidazole
Molecular Weight (g/mol) 380.44
Formula Weight 380.1736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.