missing translation for 'onlineSavingsMsg'
Learn More

Ciclopirox Ethanolamine, TRC

Product Code. 30180074
Change view
Click to view available options
Quantity:
50 mg
500 mg
5 g
Unit Size:
500mg
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180074 5 g 5g
30180197 500 mg 500mg
30180109 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180074 Supplier Toronto Research Chemicals Supplier No. TRCC4328055G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41621-49-2
Molecular Formula C12 H17 N O2 . C2 H7 N O
Molecular Weight (g/mol) 268.35
Synonym Ethanol, 2-amino-, compd. with 6-cyclohexyl-1-hydroxy-4-methyl-2(1H)-pyridinone (1:1) (9CI), 1-Hydroxy-4-methyl-6-cyclohexyl-2-pyridinone ethanolamine salt, Batrafen, Brumixol, Ciclochem, Ciclopirox ethanolamine (1:1), Ciclopirox olamine, Fungowas, HOE 296, Loprox, Micoxolamina, Mycoster, NSC 336278, Sebiprox, Selergo, Terit
IUPAC Name 2-aminoethanol;6-cyclohexyl-1-hydroxy-4-methylpyridin-2-one
SMILES CC1=CC(=O)N(O)C(=C1)C2CCCCC2.NCCO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H17NO2.C2H7NO/c1-9-7-11(13(15)12(14)8-9)10-5-3-2-4-6-10;3-1-2-4/h7-8,10,15H,2-6H2,1H3;4H,1-3H2
Formula Weight 268.1787
Chemical Name or Material Ciclopirox olamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.