missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ciclopirox olamine

Product Code. 16594234 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16594234 1 g 1g
16504244 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16594234 Supplier Thermo Scientific Chemicals Supplier No. 463440010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41621-49-2
Molecular Formula C14H24N2O3
Molecular Weight (g/mol) 268.36
InChI Key MBRHNTMUYWQHMR-UHFFFAOYSA-N
IUPAC Name 2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
SMILES NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1

Specifications

Melting Point 143.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: almost insoluble in dimethylsulfoxide and methanol
Formula Weight 268.35
Chemical Name or Material Ciclopirox olamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.