missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cidofovir dihydrate

Product Code. 16641852 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16641852 1 g 1g
16651852 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16641852 Supplier Thermo Scientific Chemicals Supplier No. 463120010
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Format Powder
Molecular Weight (g/mol) 315.22
Product Type Cidofovir dihydrate
Source synthetic source
Content And Storage Freezer -20°C
Shipping Condition normal conditions
Purity ≥97.5 % by HPLC
Quantity 1 g
Color White to almost white

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.