missing translation for 'onlineSavingsMsg'
Learn More

Ciglitazone, Tocris Bioscience™

Product Code. 18702664
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18702664 10 mg 10mg
18742634 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18702664 Supplier Tocris Bioscience Supplier No. 1307/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective PPARγ agonist

Selective agonist at PPARγ (peroxisome proliferator-activated receptor γ). Activates PPARγ with an EC50 value of 3 μM in vitro, and is at least 33-fold selective over PPARα and δ. Antihyperglycemic in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74772-77-3
Molecular Formula C18H23NO3S
Molecular Weight (g/mol) 333.446
InChI Key YZFWTZACSRHJQD-UHFFFAOYSA-N
Synonym ciglitazone, ciglitizone, ciglitazona, ciglitazonum, ciglitazonum latin, ciglitazona spanish, ciglitazone usan:inn, +--5-p-1-methylcyclohexyl methoxy benzyl-2,4-thiazolidinedione, +/--5-4-1-methylcyclohexylmethoxy benzyl thiazolidine-2,4-dione, 5-4-1-methylcyclohexyl methoxy benzyl-1,3-thiazolidine-2,4-dione
PubChem CID 2750
ChEBI CHEBI:64227
IUPAC Name 5-[[4-[(1-methylcyclohexyl)methoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
SMILES CC1(CCCCC1)COC2=CC=C(C=C2)CC3C(=O)NC(=O)S3

Specifications

Quantity 10 mg
Formula Weight 333.44
Percent Purity >99%
Chemical Name or Material Ciglitazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.