missing translation for 'onlineSavingsMsg'
Learn More

Cilastatin Ammonium Salt, TRC

Product Code. 30132786
Change view
Click to view available options
Quantity:
10 mg
100 mg
2.5 g
Unit Size:
100mg
10mg
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132786 2.5 g 2.5g
30007231 10 mg 10mg
30012877 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132786 Supplier Toronto Research Chemicals Supplier No. TRCC4411102.5G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877674-82-3
Molecular Formula C16 H25 N2 O5 S . H4 N
Molecular Weight (g/mol) 375.484
Synonym 2-Heptenoic acid, 7-[[(2R)-2-amino-2-carboxyethyl]thio]-2-[[[(1S)-2,2-dimethylcyclopropyl]carbonyl]amino]-, ammonium salt (1:1), (2Z)-, 2-Heptenoic acid, 7-[[(2R)-2-amino-2-carboxyethyl]thio]-2-[[[(1S)-2,2-dimethylcyclopropyl]carbonyl]amino]-, monoammoniu
IUPAC Name azanium;(Z)-7-[(2R)-2-amino-2-carboxyethyl]sulfanyl-2-[[(1S)-2,2-dimethylcyclopropanecarbonyl]amino]hept-2-enoate
SMILES [NH4+].CC1(C)C[C@@H]1C(=O)N\C(=C/CCCCSC[C@H](N)C(=O)O)\C(=O)[O-]

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C16H26N2O5S.H3N/c1-16(2)8-10(16)13(19)18-12(15(22)23)6-4-3-5-7-24-9-11(17)14(20)21;/h6,10-11H,3-5,7-9,17H2,1-2H3,(H,18,19)(H,20,21)(H,22,23);1H3/b12-6-;/t10-,11+;/m1./s1
Formula Weight 375.183
Percent Purity >95%
Chemical Name or Material Cilastatin Ammonium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.