missing translation for 'onlineSavingsMsg'
Learn More

Cilnidipine, MedChemExpress

Product Code. 16467286
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467286 100 mg 100mg
16453337 50 mg 50mg
16407328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16467286 Supplier Medchem Express Supplier No. HY17404
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cilnidipine is a long-acting, second-generation dihydropyridine Ca2+-channel blocker on L and N-type Ca2+ channel. Antihypertensive effects.

Cilnidipine is a long-acting, second-generation dihydropyridine Ca2+-channel blocker on L and N-type Ca2+ channel. Antihypertensive effects.

Specifications

Chemical Name or Material Cilnidipine
CAS 132203-70-4
Color Yellowish Green
Purity Grade Notes Research
Molecular Formula C27H28N2O7
Quantity 100 mg
Synonym FRC-8653
Solubility Information DMSO : ≥ 100 mg/mL (203.04 mM)
SMILES O=C(C1=C(C)NC(C)=C(C(OC/C=C/C2=CC=CC=C2)=O)C1C3=CC=CC([N+]([O-])=O)=C3)OCCOC
Molecular Weight (g/mol) 492.52
Formula Weight 492.52
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.