missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CIM 0216

Product Code. 18771598
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18771598 50 mg 50mg
18743939 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18771598 Supplier R&D Systems™ Supplier No. 5521/50
  • ONLINE EXCLUSIVE PRICE
    1278.88 €
    1144.50 € / 50mg
    Save 134.38 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective TRPM3 agonist

CIM 0216 is a selective TRPM3 agonist. Exhibits selectivity for TRPM3 over TRPM1, TRPM2 and TRPM4-8. Stimulates insulin release from pancreatic islet cells in vitro. Evokes nocifensive behavior in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide
CAS 1031496-06-6
Quantity 50 mg
Product Line Tocris
Target TRPM Agonists
Molecular Formula C21H21N3O2
Formula Weight 347.41
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.