missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CIM 0216
GREENER_CHOICE

Product Code. 18743939
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18743939

Brand: Tocris Bioscience™ 5521/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TRPM3 agonist

CIM 0216 is a selective TRPM3 agonist. Exhibits selectivity for TRPM3 over TRPM1, TRPM2 and TRPM4-8. Stimulates insulin release from pancreatic islet cells in vitro. Evokes nocifensive behavior in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dihydro-N-(5-methyl-3-isoxazolyl)-α-phenyl-1(2H)-quinolineacetamide
CAS 1031496-06-6
Quantity 10 mg
Target TRPM Agonists
Molecular Formula C21H21N3O2
Formula Weight 347.41
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.