missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cimetidine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Cimetidine |
| CAS | 51481-61-9 |
| Molecular Formula | C10 H16 N6 S |
| Quantity | 1 g |
| Synonym | Cimetidine, N-Cyano-N'-methyl-N''-[2-[[(4-methyl-1H-imidazol-5-yl)methyl]thio]ethyl]guanidine, N-Cyano-N'-methyl-N''-[2-[[(5-methyl-1H-imidazol-4-yl)methyl]thio]ethyl]guanidine, Acibilin, Acinil, Biomet, Cimal, Cimetag, Cimetum, Dyspamet, Edalene, Eureceptor, Gastromet, Histodil, N-Cyano-N'-methyl-N''-[2-((4-methyl-5-imidazolyl)-methylthio)ethyl]guanidine, NSC 335308, Peptol, SKF 92334, Tagamet, Tametin, Tratul, Ulcedin, Ulcedine, Ulcerfen, Ulcimet, Ulcofalk, Ulcomedina, Ulcomet, Ulhys |
| InChI Formula | InChI=1S/C10H16N6S/c1-8-9(16-7-15-8)5-17-4-3-13-10(12-2)14-6-11/h7H,3-5H2,1-2H3,(H,15,16)(H2,12,13,14) |
| SMILES | CNC(=NC#N)NCCSCc1nc[nH]c1C |
| IUPAC Name | 2-cyano-1-methyl-3-[2-[(5-methyl-1H-imidazol-4-yl)methylsulfanyl]ethyl]guanidine |
| Molecular Weight (g/mol) | 252.34 |
| Formula Weight | 252.1157 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction