missing translation for 'onlineSavingsMsg'
Learn More

Cinchophen, TRC

Product Code. 30364795
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30364795 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30364795 Supplier Toronto Research Chemicals Supplier No. TRCC4419155G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cinchophen
CAS 132-60-5
Molecular Formula C16 H11 N O2
Quantity 5 g
Synonym 4-Quinolinecarboxylic acid, 2-phenyl-, Cinchoninic acid, 2-phenyl- (8CI), 2-Phenyl-4-quinolinecarboxylic acid, 2-Phenylcinchonic acid, 2-Phenylcinchoninic acid, 4-Carboxy-2-phenylquinoline, Aciphenochinoline, Aciphenochinolinium, Agotan, Alutyl, Alutyo, Aminophan, Artam, Artexin, Atigoa, Atocin, Atofan, Atophan, Cinchophen, Cinchophene, Cinchophenic acid, Cinconal, Cincophen, Cincosal, Ikterosan, Mylofanol, NSC 2617, NSC 70178, Phenophan, Phenoquin, Polyphlogin, Quinofen, Quinophan, Quinophen, Rhematan, Rheumatan, Rheumin, Rheumine, Tervalon, Tophol, Tophosan, Traubofan, Vantyl, Viophan
InChI Formula InChI=1S/C16H11NO2/c18-16(19)13-10-15(11-6-2-1-3-7-11)17-14-9-5-4-8-12(13)14/h1-10H,(H,18,19)
SMILES OC(=O)c1cc(nc2ccccc12)c3ccccc3
IUPAC Name 2-phenylquinoline-4-carboxylic acid
Molecular Weight (g/mol) 249.26
Formula Weight 249.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.