missing translation for 'onlineSavingsMsg'
Learn More

Cinobufagine-d3, TRC

Product Code. 30349200
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349200 1 mg 1mg
30352501 5 mg 5mg
30366528 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30349200 Supplier Toronto Research Chemicals Supplier No. TRCC4755021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C26H31D3O6
Molecular Weight (g/mol) 445.56
Synonym NSC 90325-d3, Cinobufagin-d3, 14,15-Epoxy-14H-cyclopenta[a]phenanthrene bufa-20,22-dienolide-d3 deriv., 14,15β-Epoxy-3β,16β-dihydroxy-5β-bufa-20,22-dienolide-d3 16-Acetate, (3β,5β,15β,16β)-16-(Acetyloxy)-14,15-epoxy-3-hydroxy-bufa-20,22-dienolide-d3
IUPAC Name [(1R,2S,4R,5R,6R,7R,10S,11S,14S,16R)-14-hydroxy-7,11-dimethyl-6-(6-oxopyran-3-yl)-3-oxapentacyclo[8.8.0.02,4.02,7.011,16]octadecan-5-yl] 2,2,2-trideuterioacetate
SMILES O[C@H]1CC[C@@]2(C)[C@@](CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@]3(O5)[C@H]5[C@H](OC(C([2H])([2H])[2H])=O)[C@@H]4C(C=C6)=COC6=O)([H])C1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C26H34O6/c1-14(27)31-22-21(15-4-7-20(29)30-13-15)25(3)11-9-18-19(26(25)23(22)32-26)6-5-16-12-17(28)8-10-24(16,18)2/h4,7,13,16-19,21-23,28H,5-6,8-12H2,1-3H3/t16-,17+,18+,19-,21+,22-,23-,24+,25-,26-/m1/s1/i1D3
Chemical Name or Material Cinobufagine-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.