missing translation for 'onlineSavingsMsg'
Learn More

Cinoxacin, TRC

Product Code. 17988104
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17988104 1 g 1g
17957824 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17988104 Supplier Toronto Research Chemicals Supplier No. TRCC4757001G
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28657-80-9
Molecular Formula C12 H10 N2 O5
Molecular Weight (g/mol) 262.22
Synonym [1,3]Dioxolo[4,5-g]cinnoline-3-carboxylic acid, 1-ethyl-1,4-dihydro-4-oxo-, 1-Ethyl-3-carboxy-6,7-methylenedioxy-4-cinnolone, Cinobac, Cinobactin, Cinoxacin, Compound 64716, NSC 304467, Noxigram, Uronorm
IUPAC Name 1-ethyl-4-oxo-[1,3]dioxolo[4,5-g]cinnoline-3-carboxylic acid
SMILES CCN1N=C(C(=O)O)C(=O)c2cc3OCOc3cc12

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H10N2O5/c1-2-14-7-4-9-8(18-5-19-9)3-6(7)11(15)10(13-14)12(16)17/h3-4H,2,5H2,1H3,(H,16,17)
Formula Weight 262.059
Chemical Name or Material Cinoxacin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.