missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CINPA 1

Product Code. 18701378
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18701378

Brand: Tocris Bioscience™ 5605/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent CAR antagonist

CINPA 1 is a potent CAR antagonist (IC50 = 70 nM). Exhibits >90-fold selectivity for CAR over pregnane X receptor (PXR). Also selective over a panel of other nuclear receptors including FXR, LXR, PPARγ and RXR.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl [5-[(diethylamino)acetyl]-10,11-dihydro-5H-dibenz[b,f]azepin-3-yl]carbamate
CAS 102636-74-8
Quantity 10 mg
Target Constitutive Androstane Receptor Antagonists
Molecular Formula C23H29N3O3
Formula Weight 395.49
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.