missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin hydrochloride monohydrate, Thermo Scientific™

An anti-bacterial agent effective against anaerobic bacteria

72.10 € - 186.00 €

Chemical Identifiers

CAS 86393-32-0
Molecular Formula C17H21ClFN3O4
Molecular Weight (g/mol) 385.82
MDL Number MFCD00242856
InChI Key ARPUHYJMCVWYCZ-UHFFFAOYSA-N
PubChem CID 62998
ChEBI CHEBI:59936
IUPAC Name hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
SMILES [H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15454449
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
J61970.06
5g
72.10 €
5g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15464449
SDS View Documents
Thermo Scientific Alfa Aesar
J61970.14
25g
186.00 €
25g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ciprofloxacin hydrochloride has been suggested as a treatment for a wide range of infections caused by susceptible organisms including infections of urinary, respiratory, and gastrointestinal tracts, gonorrhoea and septicaemia. May be given by mouth or may be administered by intravenous infusion. Nalidixic acid analogue.

Solubility
Soluble in water (35mg/ml).

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications

Chemical Identifiers

86393-32-0
385.82
ARPUHYJMCVWYCZ-UHFFFAOYSA-N
CHEBI:59936
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
C17H21ClFN3O4
MFCD00242856
62998
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride

Specifications

86393-32-0
C17H21ClFN3O4
14,2314
ARPUHYJMCVWYCZ-UHFFFAOYSA-N
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
62998
385.82 (367.80 Anhydrous)
White-Yellow
MFCD00242856
Soluble in water (35mg/ml).
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
385.82
CHEBI:59936
Ciprofloxacin hydrochloride
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : VB1993800

missing translation for 'tsca' : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.