missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin hydrochloride monohydrate, Thermo Scientific™

An anti-bacterial agent effective against anaerobic bacteria

Brand:  Thermo Scientific Alfa Aesar J61970.14

Quantity 25g

 View more versions of this product

Product Code. 15464449

  • 186.00 € / 25g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ciprofloxacin hydrochloride has been suggested as a treatment for a wide range of infections caused by susceptible organisms including infections of urinary, respiratory, and gastrointestinal tracts, gonorrhoea and septicaemia. May be given by mouth or may be administered by intravenous infusion. Nalidixic acid analogue.

Solubility
Soluble in water (35mg/ml).

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

86393-32-0
385.82
ARPUHYJMCVWYCZ-UHFFFAOYSA-N
CHEBI:59936
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
C17H21ClFN3O4
MFCD00242856
62998
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride

Specifications

86393-32-0
25g
MFCD00242856
Soluble in water (35mg/ml).
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
385.82
CHEBI:59936
Ciprofloxacin hydrochloride
White-Yellow
C17H21ClFN3O4
14,2314
ARPUHYJMCVWYCZ-UHFFFAOYSA-N
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
62998
385.82 (367.80 Anhydrous)
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.