missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin Hydrochloride Monohydrate, TRC

Product Code. 30134186
Change view
Click to view available options
Quantity:
250 mg
2.5 g
10 g
Unit Size:
10g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134186 10 g 10g
30003368 2.5 g 2.5g
17933385 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134186 Supplier Toronto Research Chemicals Supplier No. TRCC48250310G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86393-32-0
Molecular Formula C17 H18 F N3 O3 . Cl H . H2 O
Molecular Weight (g/mol) 385.82
Synonym Ciprofloxacin Hydrochloride Monohydrate, Enrofloxacin Imp. B (EP) as Hydrochloride Monohydrate, 1-Cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid monohydrochloride monohydrate
IUPAC Name 1-cyclopropyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrate;hydrochloride
SMILES O.Cl.OC(=O)C1=CN(C2CC2)c3cc(N4CCNCC4)c(F)cc3C1=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H18FN3O3.ClH.H2O/c18-13-7-11-14(8-15(13)20-5-3-19-4-6-20)21(10-1-2-10)9-12(16(11)22)17(23)24;;/h7-10,19H,1-6H2,(H,23,24);1H;1H2
Formula Weight 385.1205
Percent Purity >95%
Chemical Name or Material Ciprofloxacin Hydrochloride Monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.