missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Circuit Breaker Board

Product Code. 11776695 Shop All Brady Products
Change view
Click to view available options
Color:
Red/White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11776695 Red/White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11776695 Supplier Brady™ Supplier No. 800124
  • ONLINE EXCLUSIVE PRICE
    76.30 €
    73.41 € / Pack of 1
    Save 2.89 €
    3% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Circuit Breaker Board for lockout tagout.

  • For lockout tagout.

Specifications

Product Type Lockout Station
Color Red/White
Material Plastic
Description Circuit breaker lockout Center
Displayed Text CIRCUIT BREAKER LOCKOUT EQUIPMENT
For Use With (Application) Lockout Tagout
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.