missing translation for 'onlineSavingsMsg'
Learn More

(±)-cis-1,2-Diaminocyclohexane, 97%

Product Code. 10244544 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10244544 1 g Glass bottle 1g
10778425 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10244544

Brand: Thermo Scientific Acros 248660010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1436-59-5 | C6H14N2 | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1436-59-5
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.19
MDL Number MFCD00063746
InChI Key SSJXIUAHEKJCMH-UHFFFAOYNA-N
Synonym cis-1,2-diaminocyclohexane, cis-1,2-cyclohexanediamine, 1r,2s-cyclohexane-1,2-diamine, unii-r0zn6k26ep, cis-cyclohexane-1,2-diamine, 1,2-diaminocyclohexane, cis, r0zn6k26ep, 1,2-cyclohexanediamine, 1r,2s-rel, 1,2-cyclohexanediamine #, cis-1,2-cyclohexandiamine
PubChem CID 342917
SMILES NC1CCCCC1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown
Density 0.9520g/mL
Boiling Point 92.0°C (14.0 Mbar)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4930 to 1.4960 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.952
IUPAC Name (1S,2R)-cyclohexane-1,2-diamine
Formula Weight 114.19
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material (±)-cis-1, 2-Diaminocyclohexane
Show More Show Less
compliance-icons
Product Identifier
  • (+/-)-cis-1,2-Diaminocyclohexane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.