missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Diacetoxy-2-butene, 95%

Product Code. 10676620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10676620 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10676620 Supplier Thermo Scientific Acros Supplier No. 431260250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 25260-60-0 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25260-60-0
Color Colorless
Density 1.0800g/mL
Boiling Point 222.0°C to 223.0°C
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C8H12O4
MDL Number MFCD00059339,MFCD00077968
Quantity 25 mL
Specific Gravity 1.08
Synonym cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate
InChI Key VZUAUHWZIKOMFC-ARJAWSKDSA-N
SMILES CC(=O)OC\C=C/COC(C)=O
IUPAC Name [(Z)-4-acetyloxybut-2-enyl] acetate
Molecular Weight (g/mol) 172.18
PubChem CID 643799
Formula Weight 172.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material cis-1, 4-Diacetoxy-2-butene
Show More Show Less
compliance-icons
Product Identifier
  • cis-1,4-Diacetoxy-2-butene
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.