missing translation for 'onlineSavingsMsg'
Learn More

cis-1,4-Diacetoxy-2-butene, 96%

Product Code. 11347846
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347846 50 g 50g
11337846 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347846 Supplier Thermo Scientific Alfa Aesar Supplier No. L13024.18
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25260-60-0 | C8H12O4 | 172.18 g/mol

cis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-1,4-Diacetoxy-2-butene is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25260-60-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00059339,MFCD00077968
InChI Key VZUAUHWZIKOMFC-ARJAWSKDSA-N
Synonym cis-1,4-diacetoxy-2-butene, z-but-2-ene-1,4-diyl diacetate, 2z-but-2-ene-1,4-diyl diacetate, cis-2-butene-1,4-diol diacetate, 2z-4-acetyloxybut-2-enyl acetate, 2z-4-acetyloxy but-2-en-1-yl acetate, cis-1,4-diacetoxy-2-buten, z-1,4-diacetoxy-2-butene, cis-2-buten-1,4-diol diacetate, z-but-2-ene-1,4-diyldiacetate
PubChem CID 643799
IUPAC Name [(Z)-4-acetyloxybut-2-enyl] acetate
SMILES CC(=O)OC\C=C/COC(C)=O

Specifications

Density 1.09
Boiling Point 120°C to 121°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4416
Quantity 50 g
Beilstein 1724379
Solubility Information Not miscible in water.
Formula Weight 172.18
Percent Purity 96%
Chemical Name or Material cis-1,4-Diacetoxy-2-butene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.