missing translation for 'onlineSavingsMsg'
Learn More

cis-1,5-Dimethylbicyclo[3.3.0]octane-3,7-dione, 97%, Thermo Scientific™

Product Code. 10533761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
10533761 1 g Glass bottle 1g
1 options
This item is not returnable. View return policy

Product Code. 10533761

Brand: Thermo Scientific Acros 205550010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 21170-10-5
Melting Point 228.0°C to 230.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C10H14O2
MDL Number MFCD00003772
Quantity 1 g
Synonym cis-1,5-dimethylbicyclo 3.3.0 octane-3,7-dione, 3a,6a-dimethylperhydropentalene-2,5-dione, cis-3a,6a-dimethyltetrahydropentalene-2,5 1h,3h-dione, cis-tetrahydro-3a,6a-dimethylpentalene-2,5 1h,3h-dione, 3a,6a-dimethyltetrahydro-2,5 1h,3h-pentalenedione, 3a,6a-dimethyl-tetrahydropentalene-2,5-dione, 3ar,6ar-3a,6a-dimethyl-tetrahydropentalene-2,5-dione, zlchem 111, acmc-20alfk, pubchem6298
InChI Key KGZWVKASYOPUET-UHFFFAOYSA-N
SMILES CC12CC(=O)CC1(CC(=O)C2)C
IUPAC Name 3a,6a-dimethyl-1,3,4,6-tetrahydropentalene-2,5-dione
Molecular Weight (g/mol) 166.22
PubChem CID 89460
Formula Weight 166.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material cis-1,5-Dimethylbicyclo[3.3.0]octane-3,7-dione, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • cis-1,5-Dimethylbicyclo¢3.3.0!octane-3,7-dione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.