missing translation for 'onlineSavingsMsg'
Learn More

cis-10,11-Dihydroxy-10,11-dihydrocarbamazepine, TRC

Product Code. 30007151
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007151 5 mg 5mg
30004176 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007151 Supplier Toronto Research Chemicals Supplier No. TRCD4488155MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58955-94-5
Molecular Formula C15H14N2O3
Molecular Weight (g/mol) 270.28
Synonym 5H-Dibenz[b,f]azepine-5-carboxamide, 10,11-dihydro-10,11-dihydroxy-, (10R,11S)-rel-, 5H-Dibenz[b,f]azepine-5-carboxamide, 10,11-dihydro-10,11-dihydroxy-, cis-, rel-(10R,11S)-10,11-Dihydro-10,11-dihydroxy-5H-dibenz[b,f]azepine-5-carboxamide, CGP 5924, cis-10,11-Dihydro-10,11-dihydroxycarbamazepine, cis-10,11-Dihydroxy-10,11-dihydrocarbamazepine, cis-Carbamazepine-10,11-dihydro-10,11-dihydroxide
IUPAC Name (5S,6R)-5,6-dihydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
SMILES NC(=O)N1c2ccccc2[C@H](O)[C@H](O)c3ccccc13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H14N2O3/c16-15(20)17-11-7-3-1-5-9(11)13(18)14(19)10-6-2-4-8-12(10)17/h1-8,13-14,18-19H,(H2,16,20)/t13-,14+
Formula Weight 270.1004
Percent Purity >95%
Chemical Name or Material cis-10,11-Dihydroxy-10,11-dihydrocarbamazepine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.