missing translation for 'onlineSavingsMsg'
Learn More

cis (2,3)-Dihydro Tetrabenazine, TRC

Product Code. 30384337
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384337 1 mg -
30375833 10 mg -
30369516 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384337 Supplier Toronto Research Chemicals Supplier No. TRCD4496501MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113627-25-1
Molecular Formula C19 H29 N O3
Molecular Weight (g/mol) 319.44
Synonym rel-(2R,3S,11bS)-1,3,4,6,7,11b-Hexahydro-9,10-dimethoxy-3-(2-methylpropyl)-2H-benzo[a]quinolizin-2-ol, (2α,3α,11bα)-1,3,4,6,7,11b-Hexahydro-9,10-dimethoxy-3
IUPAC Name (2R,3S,11bS)-9,10-dimethoxy-3-(2-methylpropyl)-2,3,4,6,7,11b-hexahydro-1H-benzo[a]quinolizin-2-ol
SMILES COc1cc2CCN3C[C@H](CC(C)C)[C@H](O)C[C@H]3c2cc1OC

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C19H29NO3/c1-12(2)7-14-11-20-6-5-13-8-18(22-3)19(23-4)9-15(13)16(20)10-17(14)21/h8-9,12,14,16-17,21H,5-7,10-11H2,1-4H3/t14-,16-,17+/m0/s1
Formula Weight 319.2147
Chemical Name or Material cis (2,3)-dihydro Tetrabenazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.