missing translation for 'onlineSavingsMsg'
Learn More

cis-2,3-Epoxybutane, 97%

Product Code. 10087442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10087442 1 g 1g
10033690 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 10087442

Brand: Thermo Scientific Acros 307870010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1758-33-4 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1758-33-4
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005128
InChI Key PQXKWPLDPFFDJP-ZXZARUISSA-N
Synonym cis-2,3-epoxybutane, cis-2-butene oxide, cis-2,3-dimethyloxirane, oxirane, 2,3-dimethyl-, cis, cis-2-butene epoxide, cis-2-butylene oxide, butane, 2,3-epoxy-, cis, ccris 3758, 2r,3s-2,3-dimethyloxirane, meso-2,3-epoxybutane
PubChem CID 92162
IUPAC Name (2R,3S)-2,3-dimethyloxirane
SMILES C[C@H]1O[C@H]1C

Specifications

Melting Point -84°C to 83°C
Color Brown to White or Purple
Density 0.8260g/mL
Boiling Point 60.0°C to 61.0°C
Flash Point −21.7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (corrected for H2O) (GC)
Packaging Glass bottle
Refractive Index 1.3820 to 1.3840
Quantity 1 g
Specific Gravity 0.826
Formula Weight 72.12
Percent Purity 97%
Physical Form Powder
Chemical Name or Material cis-2, 3-Epoxybutane, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.