missing translation for 'onlineSavingsMsg'
Learn More

cis-2,3-Epoxybutane, 98%, Thermo Scientific Chemicals

Product Code. 11328375
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11328375 5 g 5g
11318375 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11328375

Brand: Thermo Scientific Alfa Aesar L10320.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the enantioselective methanolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the enantioselective methanolysis.

Solubility
Soluble in water, 2.848e+004 mg/L.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Store at 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1758-33-4
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005128
InChI Key PQXKWPLDPFFDJP-ZXZARUISSA-N
Synonym cis-2,3-epoxybutane, cis-2-butene oxide, cis-2,3-dimethyloxirane, oxirane, 2,3-dimethyl-, cis, cis-2-butene epoxide, cis-2-butylene oxide, butane, 2,3-epoxy-, cis, ccris 3758, 2r,3s-2,3-dimethyloxirane, meso-2,3-epoxybutane
PubChem CID 92162
IUPAC Name (2R,3S)-2,3-dimethyloxirane
SMILES C[C@H]1O[C@H]1C

Specifications

Melting Point -84°C
Density 0.826
Boiling Point 60°C to 61°C
Flash Point −21°C (−5°F)
Refractive Index 1.383
Quantity 5 g
UN Number UN3271
Beilstein 79770
Solubility Information Soluble in water,2.848e+004mg/L.
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material cis-2,3-Epoxybutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.