missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylpiperidine, 97+%, Thermo Scientific™

Product Code. 10142670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10142670 250 g Glass bottle 250g
10316700 5 g Glass bottle 5g
10326700 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10142670 Supplier Thermo Scientific Acros Supplier No. 116682500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-17-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.2
MDL Number MFCD00005986
InChI Key SDGKUVSVPIIUCF-KNVOCYPGSA-N
Synonym cis-2,6-dimethylpiperidine, 2r,6s-2,6-dimethylpiperidine, cis-hexahydro-2,6-lutidine, cis-2,6-lupetidine, piperidin, 2e,6e-dimethyl, cis-2,6-dimethyl-piperidine, cis-lupetidine, cis-nanophine, prestwick0_000666, prestwick1_000666
PubChem CID 641768
IUPAC Name (2R,6S)-2,6-dimethylpiperidine
SMILES CC1CCCC(N1)C

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 127°C to 128°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4384 to 1.4404
Quantity 250 g
UN Number 2733
Beilstein 20, 108
Fieser 02,156
Specific Gravity 0.84
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 113.2
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material cis-2, 6-Dimethylpiperidine, 99%, min. 95% cis-isomer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-2,6-Dimethylpiperidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.