missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Amino-1-cyclopentanecarboxamide, 98%

Product Code. 10653142 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
10653142 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 10653142

Brand: Thermo Scientific Acros 334420010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135053-11-1 | C6H12N2O | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 131.0°C to 137.0°C
CAS 135053-11-1
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C6H12N2O
Packaging Glass Bottle
Linear Formula C5H8(NH2)CONH2
MDL Number MFCD01863253
Synonym cis-2-aminocyclopentanecarboxamide, cis-2-amino-1-cyclopentanecarboxamide, 1r,2s-2-aminocyclopentane-1-carboxamide, 1r,2s-2-aminocyclopentanecarboxamide, 2beta-aminocyclopentane-1beta-carboxamide, +/--cis-2-aminocyclopentanecarboxamide, cis-2-amino-cyclopentanecarboxylic acid amide, cyclopentanecarboxamide, 2-amino-, 1r,2s, 1r,2s-2-amino-cyclopentanecarboxylic acid amide, cyclopentanecarboxamide, 2-amino-, 1r,2s-9ci
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform,ethanol,dmso,,dmf,hydrochloric acid
InChI Key FUGFTUCRJJFPES-UHNVWZDZSA-N
SMILES C1CC(C(C1)N)C(=O)N
IUPAC Name (1R,2S)-2-aminocyclopentane-1-carboxamide
Molecular Weight (g/mol) 128.17
Quantity 1 g
PubChem CID 2733686
Formula Weight 128.17
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material cis-2-Amino-1-cyclopentanecarboxamide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.