missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Amino-1-cyclopentanecarboxylic acid hydrochloride hemihydrate, 99%, Thermo Scientific™

Product Code. 10257130 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
10257130 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 10257130

Brand: Thermo Scientific Acros 298382500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 199°C to 202°C
CAS 212755-84-5
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula HCl·1/2H2O
Packaging Glass Bottle
MDL Number MFCD00800502
Synonym 1s,2r-2-aminocyclopentane-1-carboxylic acid hydrate hydrochloride, cis-2-amino-1-cyclopentanecarboxylic acid hydrochloride hemihydrate, 99
Solubility Information Solubility in water: soluble in water
InChI Key LVBDVNLIEHCCTP-JBUOLDKXSA-N
SMILES C1CC(C(C1)N)C(=O)O.Cl
IUPAC Name (1R,2S)-2-aminocyclopentane-1-carboxylic acid;hydrochloride
Molecular Weight (g/mol) 174.63
Quantity 250 mg
PubChem CID 2733976
Formula Weight 174.63
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material cis-2-Amino-1-cyclopentanecarboxylic acid hydrochloride hemihydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.