missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Butene-1,4-diol, 97%

Product Code. 10085753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10085753 100 mL Glass bottle 100mL
10254610 500 mL Glass bottle 500mL
11954631 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10085753 Supplier Thermo Scientific Acros Supplier No. 107721000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6117-80-2 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6117-80-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00002924,MFCD00063207
InChI Key ORTVZLZNOYNASJ-UPHRSURJSA-N
Synonym cis-2-butene-1,4-diol, z-2-butene-1,4-diol, cis-butenediol, unii-za7vgu6scv, cis-1,4-dihydroxy-2-butene, z-but-2-ene-1,4-diol, 2-butene-1,4-diol, 2z-but-2-ene-1,4-diol, 2-butene-1,4-diol, z, za7vgu6scv
PubChem CID 643790
SMILES OC\C=C/CO

Specifications

Melting Point 7.0°C
Color Colorless
Density 1.0700g/mL
Boiling Point 235.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (cis-isomer) (GC)
Packaging Glass bottle
Linear Formula HOCH2CH=CHCH2OH
Refractive Index 1.4770 to 1.4800
Quantity 100 mL
Specific Gravity 1.07
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene
IUPAC Name (Z)-but-2-ene-1,4-diol
Formula Weight 88.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material cis-2-Butene-1, 4-diol, 97% cis
Show More Show Less
compliance-icons
Product Identifier
  • cis-2-Butene-1,4-diol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.