missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Chloroacrylic acid, 98+%

Product Code. 11442537
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442537 5 g 5g
11432537 1 g 1g
11452537 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442537 Supplier Thermo Scientific Alfa Aesar Supplier No. A14798.06
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1609-93-4 | C3H3ClO2 | 106.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609-93-4
Molecular Formula C3H3ClO2
Molecular Weight (g/mol) 106.51
MDL Number MFCD00004368
InChI Key MHMUCYJKZUZMNJ-UPHRSURJSA-N
Synonym cis-3-chloroacrylic acid, z-3-chloroacrylic acid, cis-beta-chloroacrylic acid, 2-propenoic acid, 3-chloro-, z, cis-3-chloropropenoic acid, 2z-3-chloroacrylic acid, 2-propenoic acid, 3-chloro-, 2z, cis-.beta.-chloroacrylic acid, z-3-chloroprop-2-enoic acid, acrylic acid, 3-chloro-, z-8ci
PubChem CID 643794
ChEBI CHEBI:27397
IUPAC Name (Z)-3-chloroprop-2-enoic acid
SMILES OC(=O)\C=C/Cl

Specifications

Melting Point 60°C to 62°C
Quantity 5 g
UN Number UN3261
Beilstein 1720278
Formula Weight 106.51
Percent Purity ≥98%
Chemical Name or Material cis-3-Chloroacrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.