missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hepten-1-ol, 97%

Product Code. 11449456
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449456 25 g 25g
11439456 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449456 Supplier Thermo Scientific Alfa Aesar Supplier No. A19172.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1708-81-2 | C7H14O | 114.19 g/mol

cis-3-Hepten-1-ol is used in the analysis of hop-derived terpenoids in beer. Imparts a fresh flower garden odor so used in flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-3-Hepten-1-ol is used in the analysis of hop-derived terpenoids in beer. Imparts a fresh flower garden odor so used in flavoring agent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1708-81-2
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00274760
InChI Key SDZQUCJFTUULJX-PLNGDYQASA-N
Synonym cis-3-hepten-1-ol, z-hept-3-en-1-ol, 3-hepten-1-ol, z, 3z-hept-3-en-1-ol, unii-lb4d00v63h, z-3-heptenol, cis-3-heptenol, z-3-hepten-1-ol, 3z-3-hepten-1-ol #
PubChem CID 5364517
IUPAC Name (Z)-hept-3-en-1-ol
SMILES CCC\C=C/CCO

Specifications

Density 0.85
Boiling Point 92°C (60 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.444
Quantity 25 g
Beilstein 1719803
Solubility Information Insoluble in water.
Formula Weight 114.18
Percent Purity 97%
Chemical Name or Material cis-3-Hepten-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.