missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Hexen-1-ol, 98%

Product Code. 10540381 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10540381 10 mL Glass bottle 10mL
10589443 50 mL Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10540381 Supplier Thermo Scientific Acros Supplier No. 120770100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 928-96-1 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-96-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00063217
InChI Key UFLHIIWVXFIJGU-ARJAWSKDSA-N
Synonym z-hex-3-en-1-ol, cis-3-hexen-1-ol, cis-3-hexenol, leaf alcohol, blatteralkohol, z-3-hexenol, z-3-hexen-1-ol, 3-hexenol, 3z-hex-3-en-1-ol, 3-hexen-1-ol, z
PubChem CID 5281167
ChEBI CHEBI:28857
SMILES CC\C=C/CCO

Specifications

Color Colorless to Yellow
Density 0.8480g/mL
Boiling Point 156.0°C to 157.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH=CHCH2CH2OH
Refractive Index 1.4380 to 1.4410
Quantity 10 mL
Beilstein 01, I, 229
Merck Index 10, 4592
Specific Gravity 0.848
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and propylene glycol, miscible with oils
IUPAC Name (3Z)-hex-3-en-1-ol
Formula Weight 100.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material cis-3-Hexen-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-3-Hexen-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.